siRNA Details
Gene Symbol: DNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080449.2 | NP_001073918.2 | 8 | 1199 | ||
NR_102264.1 | 9 | 1179 | ||||
XM_006717680.3 | XP_006717743.1 | 9 | 1579 | |||
XM_011539417.1 | XP_011537719.1 | 2 | 123 | |||
XM_017015799.1 | XP_016871288.1 | 5 | 588 | |||
GenBank mRNA | AK308333.1 | - | 8 | 1199 | ||
AK308358.1 | - | 6 | 840 | |||
BC063664.1 | - | 8 | 1112 | |||
BC111740.1 | - | 9 | 1179 | 40038149 | ||
D42046.1 | - | 8 | 1141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
