siRNA Details
Gene Symbol: DNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080449.2 | NP_001073918.2 | 19 | 3024 | ||
NR_102264.1 | 20 | 2889 | ||||
XM_006717680.3 | XP_006717743.1 | 20 | 3404 | |||
XM_011539417.1 | XP_011537719.1 | 13 | 1948 | |||
XM_017015799.1 | XP_016871288.1 | 16 | 2413 | |||
GenBank mRNA | AK307939.1 | - | 8 | 974 | ||
BC028188.1 | - | 6 | 890 | |||
BC041115.2 | - | 7 | 1042 | |||
BC053574.1 | - | 9 | 1218 | |||
BC063664.1 | - | 15 | 2223 | |||
BC073945.1 | - | 5 | 631 | |||
BC111740.1 | - | 20 | 2889 | 40038149 | ||
D42046.1 | - | 19 | 2966 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
