siRNA Details
Gene Symbol: TMEM261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318058.1 | NP_001304987.1 | 2 | 571 | ||
NM_001318059.1 | NP_001304988.1 | 2 | 473 | |||
NM_033428.2 | NP_219500.1 | 2 | 559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
