siRNA Details
Gene Symbol: TMEM261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318058.1 | NP_001304987.1 | 2 | 542 | ||
NM_001318059.1 | NP_001304988.1 | 2 | 444 | |||
NM_033428.2 | NP_219500.1 | 2 | 530 | |||
GenBank mRNA | AK128876.1 | - | 2 | 415 | ||
AK292632.1 | - | 2 | 502 | |||
AL137489.1 | - | 2 | 334 | |||
BC009510.2 | - | 2 | 503 | 3535614 | ||
BI550091.1 | - | 2 | 530 | |||
DA887295.1 | - | 2 | 530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
