siRNA Details
Gene Symbol: PLEKHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029884.1 | NP_001025055.1 | 11 | 1550 | ||
XM_005267064.3 | XP_005267121.1 | 10 | 3629 | |||
XM_005267065.3 | XP_005267122.1 | 12 | 1576 | |||
XM_005267066.2 | XP_005267123.1 | 10 | 2433 | |||
XM_006715521.1 | XP_006715584.1 | 12 | 1803 | |||
XM_006715522.3 | XP_006715585.1 | 10 | 3670 | |||
XM_011535981.2 | XP_011534283.1 | 10 | 4426 | |||
XM_011535982.2 | XP_011534284.1 | 10 | 4426 | |||
XM_011535983.2 | XP_011534285.1 | 11 | 2851 | |||
XM_017011102.1 | XP_016866591.1 | 10 | 2435 | |||
GenBank mRNA | AB033035.1 | - | 4 | 423 | ||
AJ420468.1 | - | 8 | 788 | |||
AK056300.1 | - | 10 | 1449 | |||
AK293288.1 | - | 10 | 1481 | |||
AK294798.1 | - | 10 | 1604 | |||
AK307631.1 | - | 10 | 1503 | |||
BC089428.1 | - | 6 | 661 | |||
BC140864.1 | - | 9 | 1287 | 9021732 | ||
BC151134.1 | - | 9 | 1287 | 9057034 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
