siRNA Details
Gene Symbol: PLEKHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029884.1 | NP_001025055.1 | 14 | 1740 | ||
XM_005267064.3 | XP_005267121.1 | 13 | 3819 | |||
XM_005267065.3 | XP_005267122.1 | 15 | 1766 | |||
XM_005267066.2 | XP_005267123.1 | 13 | 2623 | |||
XM_006715521.1 | XP_006715584.1 | 15 | 1993 | |||
XM_006715522.3 | XP_006715585.1 | 13 | 3860 | |||
XM_011535981.2 | XP_011534283.1 | 13 | 4616 | |||
XM_011535983.2 | XP_011534285.1 | 14 | 3041 | |||
GenBank mRNA | AB033035.1 | - | 7 | 613 | ||
AJ420468.1 | - | 11 | 978 | |||
AK056300.1 | - | 13 | 1639 | |||
AK293288.1 | - | 13 | 1671 | |||
AK294798.1 | - | 13 | 1794 | |||
AK307631.1 | - | 13 | 1693 | |||
BC089428.1 | - | 9 | 851 | |||
BC140864.1 | - | 12 | 1477 | 9021732 | ||
BC151134.1 | - | 12 | 1477 | 9057034 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
