siRNA Details
Gene Symbol: PLEKHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029884.1 | NP_001025055.1 | 17 | 6788 | ||
XM_005267064.3 | XP_005267121.1 | 16 | 8867 | |||
XM_005267065.3 | XP_005267122.1 | 18 | 6814 | |||
XM_005267066.2 | XP_005267123.1 | 16 | 7671 | |||
XM_006715521.1 | XP_006715584.1 | 18 | 7041 | |||
XM_006715522.3 | XP_006715585.1 | 16 | 8908 | |||
XM_011535981.2 | XP_011534283.1 | 16 | 9664 | |||
XM_011535982.2 | XP_011534284.1 | 15 | 9625 | |||
XM_011535983.2 | XP_011534285.1 | 17 | 8089 | |||
XM_017011102.1 | XP_016866591.1 | 15 | 7634 | |||
GenBank mRNA | AB033035.1 | - | 10 | 5661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
