siRNA Details
Gene Symbol: FUBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003934.1 | NP_003925.1 | 5 | 420 | ||
XM_005272232.1 | XP_005272289.1 | 5 | 426 | |||
XM_006717313.2 | XP_006717376.1 | 6 | 1271 | |||
XM_011519172.2 | XP_011517474.1 | 5 | 426 | |||
XM_011519173.2 | XP_011517475.1 | 3 | 678 | |||
XM_011519174.2 | XP_011517476.1 | 7 | 1516 | |||
XM_017015269.1 | XP_016870758.1 | 6 | 1271 | |||
XM_017015270.1 | XP_016870759.1 | 6 | 1271 | |||
XM_017015271.1 | XP_016870760.1 | 7 | 1516 | |||
XM_017015272.1 | XP_016870761.1 | 4 | 299 | |||
XM_017015273.1 | XP_016870762.1 | 6 | 1271 | |||
GenBank mRNA | AK295155.1 | - | 5 | 420 | ||
AL537190.3 | - | 5 | 391 | |||
BC007874.2 | - | 6 | 490 | |||
BC137338.1 | - | 5 | 417 | 9021336 | ||
BC137340.1 | - | 5 | 417 | 9021338 | ||
BI461266.1 | - | 5 | 424 | |||
U69127.1 | - | 5 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
