siRNA Details
Gene Symbol: ZBTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123329.1 | NP_001116801.1 | 2 | 1884 | ||
NM_014950.2 | NP_055765.2 | 2 | 1884 | |||
XM_005267410.1 | XP_005267467.1 | 4 | 2020 | |||
XM_011536566.2 | XP_011534868.1 | 3 | 1757 | |||
XM_011536568.2 | XP_011534870.1 | 4 | 2020 | |||
XM_017021094.1 | XP_016876583.1 | 4 | 2896 | |||
XM_017021095.1 | XP_016876584.1 | 3 | 1812 | |||
GenBank mRNA | AB023214.1 | - | 2 | 1755 | ||
AK291743.1 | - | 2 | 1662 | |||
AY598321.1 | - | 1 | 1493 | |||
BC050719.1 | - | 4 | 1924 | 6141266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
