siRNA Details
Gene Symbol: IRAK1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010844.3 | NP_001010844.1 | 4 | 671 | ||
XM_006715336.3 | XP_006715399.1 | 4 | 712 | |||
XM_011535447.1 | XP_011533749.1 | 4 | 711 | |||
XM_017010264.1 | XP_016865753.1 | 4 | 490 | |||
GenBank mRNA | AK098678.1 | - | 4 | 1168 | ||
BC112253.1 | - | 4 | 660 | 8327721 | ||
BC112255.1 | - | 4 | 660 | 8327723 | ||
BX103697.1 | - | 2 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
