siRNA Details
Gene Symbol: IRAK1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006715336.3 | XP_006715399.1 | 5 | 890 | ||
XM_011535448.2 | XP_011533750.1 | 3 | 541 | |||
XM_017010264.1 | XP_016865753.1 | 5 | 668 | |||
GenBank mRNA | AK098678.1 | - | 5 | 1346 | ||
AK309003.1 | - | 4 | 550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
