siRNA Details
Gene Symbol: FTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080432.2 | NP_001073901.1 | 9 | 4006 | ||
XM_011523313.2 | XP_011521615.1 | 9 | 4000 | |||
GenBank mRNA | AB051539.1 | - | 9 | 3788 | ||
AK096554.1 | - | 9 | 3811 | |||
BC001284.1 | - | 1 | 1060 | |||
BC003583.1 | - | 3 | 2515 | |||
BC030798.1 | - | 3 | 2772 | |||
U79260.1 | - | 1 | 1123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
