siRNA Details
Gene Symbol: HEATR5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019024.2 | NP_061897.1 | 27 | 4365 | ||
XM_006712034.2 | XP_006712097.1 | 27 | 4304 | |||
XM_006712035.3 | XP_006712098.1 | 27 | 4368 | |||
XM_011532934.2 | XP_011531236.1 | 27 | 4832 | |||
XM_011532935.2 | XP_011531237.1 | 16 | 2504 | |||
XM_017004378.1 | XP_016859867.1 | 27 | 4304 | |||
XM_017004379.1 | XP_016859868.1 | 28 | 4742 | |||
XM_017004380.1 | XP_016859869.1 | 28 | 4515 | |||
GenBank mRNA | AB037835.1 | - | 18 | 2812 | ||
AK001513.1 | - | 1 | 72 | |||
AK026928.1 | - | 4 | 625 | |||
AK096637.1 | - | 27 | 4304 | |||
BC085607.1 | - | 1 | 137 | |||
BC150605.1 | - | 27 | 4350 | 9056975 | ||
BX538008.1 | - | 21 | 3288 | |||
BX648805.1 | - | 13 | 2525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
