siRNA Details
Gene Symbol: C10orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022153.1 | NP_071436.1 | 7 | 1326 | ||
GenBank mRNA | AF193048.1 | - | 2 | 465 | ||
AF289594.1 | - | 1762 | ||||
AK024449.1 | - | 7 | 1244 | |||
AY265805.1 | - | 7 | 1241 | |||
AY358379.1 | - | 7 | 1326 | |||
BC020568.1 | - | 7 | 1198 | 4158410 | ||
BC111048.1 | - | 7 | 1174 | |||
EF506490.1 | - | 7 | 1238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
