siRNA Details
Gene Symbol: GRAMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015124.4 | NP_055939.1 | 2 | 285 | ||
XM_005261396.3 | XP_005261453.1 | 2 | 221 | |||
XM_006724169.1 | XP_006724232.1 | 2 | 183 | |||
XM_006724170.3 | XP_006724233.1 | 2 | 202 | |||
XM_011530011.1 | XP_011528313.1 | 1 | 121 | |||
XM_011530012.1 | XP_011528314.1 | 1 | 121 | |||
XM_011530013.2 | XP_011528315.2 | 2 | 230 | |||
XM_011530014.2 | XP_011528316.1 | 1 | 121 | |||
XM_017028670.1 | XP_016884159.1 | 2 | 243 | |||
GenBank mRNA | AB018310.2 | - | 2 | 285 | ||
BC129837.1 | - | 1 | 106 | 40119070 | ||
CR456470.1 | - | 1 | 111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
