siRNA Details
Gene Symbol: GRAMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015124.4 | NP_055939.1 | 19 | 4258 | ||
XM_005261396.3 | XP_005261453.1 | 19 | 4194 | |||
XM_006724169.1 | XP_006724232.1 | 19 | 4156 | |||
XM_006724170.3 | XP_006724233.1 | 19 | 4175 | |||
XM_011530011.1 | XP_011528313.1 | 18 | 3665 | |||
XM_011530012.1 | XP_011528314.1 | 17 | 4025 | |||
XM_017028670.1 | XP_016884159.1 | 19 | 4216 | |||
GenBank mRNA | BC011984.2 | - | 1 | 901 | ||
BC063558.1 | - | 1 | 849 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
