siRNA Details
Gene Symbol: ZNF37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007094.2 | NP_001007095.1 | 8 | 2009 | ||
NM_001178101.1 | NP_001171572.1 | 7 | 1531 | |||
NM_001324245.1 | NP_001311174.1 | 7 | 1460 | |||
NM_001324246.1 | NP_001311175.1 | 8 | 2005 | |||
NM_001324247.1 | NP_001311176.1 | 8 | 1750 | |||
NM_001324248.1 | NP_001311177.1 | 7 | 1715 | |||
NM_001324249.1 | NP_001311178.1 | 7 | 1711 | |||
NM_001324250.1 | NP_001311179.1 | 8 | 1825 | |||
NM_001324251.1 | NP_001311180.1 | 8 | 1829 | |||
NM_001324259.1 | NP_001311188.1 | 8 | 2059 | |||
NM_001324260.1 | NP_001311189.1 | 7 | 1581 | |||
NM_003421.2 | NP_003412.1 | 7 | 1535 | |||
XM_005252586.3 | XP_005252643.1 | 2 | 1136 | |||
XM_011519656.2 | XP_011517958.1 | 8 | 1917 | |||
XM_011519657.2 | XP_011517959.1 | 8 | 1739 | |||
XM_011519658.2 | XP_011517960.1 | 7 | 1457 | |||
XM_017016618.1 | XP_016872107.1 | 8 | 2054 | |||
XM_017016619.1 | XP_016872108.1 | 7 | 1590 | |||
XM_017016620.1 | XP_016872109.1 | 8 | 2058 | |||
XM_017016622.1 | XP_016872111.1 | 8 | 2054 | |||
GenBank mRNA | AJ492195.1 | - | 7 | 1499 | ||
AK092012.1 | - | 7 | 1524 | |||
BC015858.1 | - | 7 | 1490 | 4671816 | ||
BX647089.1 | - | 8 | 1994 | |||
X69115.1 | - | 1 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
