siRNA Details
Gene Symbol: EIF2S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001415.3 | NP_001406.1 | 12 | 1439 | ||
GenBank mRNA | AK222577.1 | - | 12 | 1431 | ||
AK290397.1 | - | 12 | 1431 | |||
BC019906.1 | - | 12 | 1439 | 4419438 | ||
L19161.1 | - | 12 | 1418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
