siRNA Details
Gene Symbol: CCDC107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195200.1 | NP_001182129.1 | 6 | 832 | ||
NM_001195201.1 | NP_001182130.1 | 6 | 812 | |||
NM_001195217.1 | NP_001182146.1 | 5 | 885 | |||
NM_174923.2 | NP_777583.2 | 5 | 913 | |||
XM_005251403.4 | XP_005251460.1 | 5 | 896 | |||
GenBank mRNA | AK075523.1 | - | 5 | 871 | ||
BC018758.1 | - | 5 | 889 | 4907976 | ||
BC051285.1 | - | 6 | 799 | 6586677 | ||
DQ884403.1 | - | 6 | 742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
