siRNA Details
Gene Symbol: MLST8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199173.1 | NP_001186102.1 | 4 | 555 | ||
NM_001199174.1 | NP_001186103.1 | 4 | 394 | |||
NM_001199175.1 | NP_001186104.1 | 4 | 582 | |||
NM_022372.4 | NP_071767.3 | 4 | 585 | |||
XM_005255475.3 | XP_005255532.2 | 4 | 361 | |||
XM_005255478.2 | XP_005255535.1 | 4 | 566 | |||
XM_005255479.2 | XP_005255536.1 | 4 | 493 | |||
XM_017023548.1 | XP_016879037.1 | 4 | 550 | |||
XM_017023549.1 | XP_016879038.1 | 4 | 291 | |||
XM_017023550.1 | XP_016879039.1 | 4 | 552 | |||
XM_017023551.1 | XP_016879040.1 | 4 | 308 | |||
XM_017023552.1 | XP_016879041.1 | 4 | 576 | |||
GenBank mRNA | AK021536.1 | - | 4 | 554 | ||
AK022227.1 | - | 4 | 364 | |||
AK098762.1 | - | 4 | 490 | |||
AK302201.1 | - | 3 | 394 | |||
AK304390.1 | - | 4 | 673 | |||
AU117583.1 | - | 4 | 554 | |||
AY223837.1 | - | 3 | 395 | |||
BC001313.2 | - | 4 | 275 | 3454025 | ||
BC017119.1 | - | 3 | 286 | 3910883 | ||
BC052292.1 | - | 3 | 809 | 5926090 | ||
BC088354.1 | - | 4 | 303 | 6168345 | ||
BI761765.1 | - | 4 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
