siRNA Details
Gene Symbol: MLST8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199173.1 | NP_001186102.1 | 4 | 573 | ||
NM_001199174.1 | NP_001186103.1 | 4 | 412 | |||
NM_001199175.1 | NP_001186104.1 | 4 | 600 | |||
NM_022372.4 | NP_071767.3 | 4 | 603 | |||
XM_005255475.3 | XP_005255532.2 | 4 | 379 | |||
XM_005255478.2 | XP_005255535.1 | 4 | 584 | |||
XM_005255479.2 | XP_005255536.1 | 4 | 511 | |||
XM_017023548.1 | XP_016879037.1 | 4 | 568 | |||
XM_017023549.1 | XP_016879038.1 | 4 | 309 | |||
XM_017023550.1 | XP_016879039.1 | 4 | 570 | |||
XM_017023551.1 | XP_016879040.1 | 4 | 326 | |||
XM_017023552.1 | XP_016879041.1 | 4 | 594 | |||
GenBank mRNA | AK021536.1 | - | 4 | 572 | ||
AK022227.1 | - | 4 | 382 | |||
AK302201.1 | - | 3 | 412 | |||
AK304390.1 | - | 4 | 691 | |||
AU117583.1 | - | 4 | 572 | |||
AU121635.1 | - | 4 | 382 | |||
AY223837.1 | - | 3 | 413 | |||
BC001313.2 | - | 4 | 293 | 3454025 | ||
BC017119.1 | - | 3 | 304 | 3910883 | ||
BC052292.1 | - | 3 | 827 | 5926090 | ||
BC088354.1 | - | 4 | 321 | 6168345 | ||
BI761765.1 | - | 4 | 331 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001252465, NM_019988, NM_001252463, NM_001252464, AK088951, and others.
Gene Ontology Categories:
