siRNA Details
Gene Symbol: LOC728024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003671.2 | 1 | 704 | |||
GenBank mRNA | CR977513.1 | - | 1 | 611 |
Gene Symbol: CXorf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170569.1 | NP_001164040.1 | 7 | 823 | ||
NM_001170570.1 | NP_001164041.1 | 6 | 699 | |||
NM_022101.3 | NP_071384.1 | 7 | 741 | |||
GenBank mRNA | AK096297.1 | - | 7 | 787 | ||
AK298877.1 | - | 6 | 677 | |||
BC023506.2 | - | 7 | 702 | |||
BQ223940.1 | - | 7 | 706 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
