siRNA Details
Gene Symbol: LMNTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173573.2 | NP_775844.2 | 6 | 657 | ||
XM_011519965.2 | XP_011518267.2 | 6 | 832 | |||
XM_011519967.2 | XP_011518269.2 | 6 | 832 | |||
XM_017017476.1 | XP_016872965.1 | 6 | 832 | |||
XM_017017477.1 | XP_016872966.1 | 6 | 817 | |||
XM_017017478.1 | XP_016872967.1 | 6 | 673 | |||
XM_017017479.1 | XP_016872968.1 | 6 | 832 | |||
GenBank mRNA | BC039077.1 | - | 6 | 657 | 5168905 | |
BX384085.2 | - | 7 | 760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
