siRNA Details
Gene Symbol: LMNTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173573.2 | NP_775844.2 | 13 | 1826 | ||
XM_011519965.2 | XP_011518267.2 | 13 | 2001 | |||
XM_011519967.2 | XP_011518269.2 | 12 | 1881 | |||
XM_017017476.1 | XP_016872965.1 | 12 | 2046 | |||
XM_017017477.1 | XP_016872966.1 | 12 | 2031 | |||
XM_017017478.1 | XP_016872967.1 | 12 | 1887 | |||
GenBank mRNA | BC037936.2 | - | 3 | 965 | ||
BC039077.1 | - | 13 | 1826 | 5168905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
