siRNA Details
Gene Symbol: KAZN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201628.2 | NP_963922.2 | 15 | 3171 | ||
XM_005245795.4 | XP_005245852.1 | 17 | 3426 | |||
XM_011541074.2 | XP_011539376.1 | 16 | 5362 | |||
XM_011541075.2 | XP_011539377.1 | 14 | 5018 | |||
XM_011541076.2 | XP_011539378.1 | 14 | 5163 | |||
XM_011541077.2 | XP_011539379.1 | 14 | 6142 | |||
XM_011541081.2 | XP_011539383.1 | 8 | 3842 | |||
GenBank mRNA | AK125388.1 | - | 1 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
