siRNA Details
Gene Symbol: RPS4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007.4 | NP_000998.1 | 7 | 914 | ||
GenBank mRNA | AK097914.1 | - | 2715 | |||
AK222479.1 | - | 7 | 843 | |||
BC000472.2 | - | 6 | 826 | 2961540 | ||
BC007308.1 | - | 6 | 758 | |||
BC071662.1 | - | 6 | 855 | 5505465 | ||
M22146.1 | - | 6 | 823 | |||
M58458.1 | - | 7 | 853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
