siRNA Details
Gene Symbol: HNRNPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318186.1 | NP_001305115.1 | 3 | 255 | ||
NM_001318187.1 | NP_001305116.1 | 3 | 255 | |||
NM_001318188.1 | NP_001305117.1 | 3 | 255 | |||
NM_002140.4 | NP_002131.2 | 3 | 416 | |||
NM_031262.3 | NP_112552.1 | 3 | 416 | |||
NM_031263.3 | NP_112553.1 | 3 | 255 | |||
XM_005251960.2 | XP_005252017.1 | 3 | 234 | |||
XM_005251963.3 | XP_005252020.1 | 3 | 268 | |||
XM_005251965.2 | XP_005252022.1 | 3 | 226 | |||
XM_011518616.1 | XP_011516918.1 | 3 | 268 | |||
XM_017014668.1 | XP_016870157.1 | 3 | 234 | |||
XM_017014669.1 | XP_016870158.1 | 3 | 226 | |||
GenBank mRNA | AB209562.1 | - | 3 | 184 | ||
AB451263.1 | - | 1 | 29 | |||
AB451390.1 | - | 1 | 29 | |||
AK096385.1 | - | 3 | 192 | |||
AK291336.1 | - | 3 | 238 | |||
AK294067.1 | - | 3 | 228 | |||
AK300744.1 | - | 3 | 198 | |||
AY911506.1 | - | 2 | 92 | |||
BC000355.2 | - | 2 | 151 | 2964383 | ||
BC014980.1 | - | 3 | 222 | 4906241 | ||
BC025321.1 | - | 3 | 174 | |||
CR456771.1 | - | 1 | 29 | |||
DA394419.1 | - | 3 | 253 | |||
DC318847.1 | - | 3 | 255 | |||
DC407458.1 | - | 3 | 288 | |||
DC428989.1 | - | 3 | 416 | |||
S74678.1 | - | 2 | 136 | |||
X72727.1 | - | 3 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
