siRNA Details
Gene Symbol: CASC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138423.3 | NP_612432.2 | 10 | 1647 | ||
NM_177974.2 | NP_816929.1 | 9 | 1479 | |||
GenBank mRNA | AK095068.1 | - | 5 | 519 | ||
AK291026.1 | - | 9 | 1236 | |||
AK298564.1 | - | 11 | 1362 | |||
AL832333.1 | - | 10 | 1419 | |||
AL832840.1 | - | 10 | 1089 | |||
AY358086.1 | - | 9 | 1430 | |||
BC041786.1 | - | 4 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
