siRNA Details
Gene Symbol: CASC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138423.3 | NP_612432.2 | 10 | 2571 | ||
NM_177974.2 | NP_816929.1 | 9 | 2403 | |||
GenBank mRNA | AF103804.1 | - | 1 | 612 | ||
AK022565.1 | - | 1 | 754 | |||
AK095068.1 | - | 5 | 1444 | |||
AL832333.1 | - | 10 | 2345 | |||
AL832840.1 | - | 10 | 2013 | |||
AY358086.1 | - | 9 | 2353 | |||
BC041786.1 | - | 4 | 1448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
