siRNA Details
Gene Symbol: CCDC107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195200.1 | NP_001182129.1 | 1 | 168 | ||
NM_001195201.1 | NP_001182130.1 | 1 | 168 | |||
NM_001195217.1 | NP_001182146.1 | 1 | 168 | |||
NM_174923.2 | NP_777583.2 | 1 | 168 | |||
XM_005251403.4 | XP_005251460.1 | 1 | 176 | |||
GenBank mRNA | AK075523.1 | - | 1 | 154 | ||
AK307223.1 | - | 1 | 93 | |||
BC018758.1 | - | 1 | 144 | 4907976 | ||
BC051285.1 | - | 1 | 155 | 6586677 | ||
BE903559.1 | - | 1 | 158 | |||
DC410699.1 | - | 1 | 168 | |||
DQ884403.1 | - | 1 | 78 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
