siRNA Details
Gene Symbol: LOC105375355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250613.4 | XP_005250670.2 | 3, 4 | 770 | ||
XM_017012885.1 | XP_016868374.1 | 3, 4 | 770 | |||
XM_017012886.1 | XP_016868375.1 | 3, 4 | 770 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 6, 7 | 1096 | |||
GenBank mRNA | BM709868.1 | - | 411 | |||
DR007973.1 | - | 1, 2 | 260 | |||
U38980.1 | - | 6, 7 | 982 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
