siRNA Details
Gene Symbol: SMIM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136503.1 | NP_001129975.1 | 2 | 218 | ||
GenBank mRNA | AF161441.1 | - | 1 | 77 | ||
BC146979.1 | - | 2 | 206 | 9056907 | ||
BC146984.1 | - | 2 | 206 | 9056912 | ||
BM769757.1 | - | 2 | 201 | |||
CB178479.1 | - | 2 | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
