siRNA Details
Gene Symbol: ARMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267041.2 | NP_001253970.1 | 15 | 1624 | ||
NM_001267042.2 | NP_001253971.1 | 15 | 1516 | |||
NM_001282342.1 | NP_001269271.1 | 14 | 1547 | |||
NM_015396.5 | NP_056211.2 | 17 | 1793 | |||
XM_006713564.3 | XP_006713627.1 | 16 | 1712 | |||
XM_006713565.2 | XP_006713628.1 | 17 | 1753 | |||
XM_006713566.3 | XP_006713629.1 | 15 | 1607 | |||
XM_006713567.3 | XP_006713630.1 | 16 | 1712 | |||
XM_011512632.1 | XP_011510934.1 | 16 | 1660 | |||
XM_011512633.1 | XP_011510935.1 | 16 | 1714 | |||
XM_017006095.1 | XP_016861584.1 | 16 | 1514 | |||
XM_017006096.1 | XP_016861585.1 | 15 | 1607 | |||
XM_017006097.1 | XP_016861586.1 | 15 | 1460 | |||
XM_017006098.1 | XP_016861587.1 | 14 | 1406 | |||
XM_017006099.1 | XP_016861588.1 | 15 | 1497 | |||
XM_017006100.1 | XP_016861589.1 | 16 | 1712 | |||
GenBank mRNA | AK001721.1 | - | 15 | 1355 | ||
AK001882.1 | - | 8 | 928 | |||
AK296757.1 | - | 15 | 1247 | |||
AK296837.1 | - | 15 | 1324 | |||
AK299229.1 | - | 11 | 985 | |||
AK299769.1 | - | 14 | 1267 | |||
AK304558.1 | - | 15 | 1300 | |||
AK315932.1 | - | 11 | 990 | |||
AL096748.1 | - | 3 | 814 | |||
BC032661.1 | - | 17 | 1657 | 5530425 | ||
BC041699.1 | - | 17 | 1472 | 5240423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
