siRNA Details
Gene Symbol: AGFG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006076.4 | NP_006067.3 | 12 | 4682 | ||
XM_005250306.2 | XP_005250363.1 | 13 | 4701 | |||
XM_017012112.1 | XP_016867601.1 | 12 | 4623 | |||
XM_017012114.1 | XP_016867603.1 | 11 | 4381 | |||
GenBank mRNA | AK057758.1 | - | 1 | 1235 | ||
AK090744.1 | - | 1 | 2448 | |||
BC047227.1 | - | 1 | 2231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
