siRNA Details
Gene Symbol: TRIM16L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037330.1 | NP_001032407.1 | 5 | 1131 | ||
XM_005256479.4 | XP_005256536.1 | 8 | 1305 | |||
XM_005256480.4 | XP_005256537.1 | 9 | 1409 | |||
XM_005256481.4 | XP_005256538.1 | 11 | 1658 | |||
XM_005256483.4 | XP_005256540.1 | 10 | 1550 | |||
XM_011523689.2 | XP_011521991.1 | 6 | 1010 | |||
XM_011523690.2 | XP_011521992.1 | 12 | 1720 | |||
XM_017024245.1 | XP_016879734.1 | 13 | 2020 | |||
XM_017024246.1 | XP_016879735.1 | 12 | 1882 | |||
XM_017024247.1 | XP_016879736.1 | 13 | 2023 | |||
XM_017024248.1 | XP_016879737.1 | 12 | 1912 | |||
XM_017024249.1 | XP_016879738.1 | 11 | 1774 | |||
XM_017024250.1 | XP_016879739.1 | 11 | 1775 | |||
XM_017024251.1 | XP_016879740.1 | 11 | 1612 | |||
XM_017024252.1 | XP_016879741.1 | 6 | 1266 | |||
GenBank mRNA | AK293428.1 | - | 9 | 1409 | ||
AK298836.1 | - | 5 | 1076 | |||
AK301639.1 | - | 3 | 839 | |||
AK301647.1 | - | 5 | 1197 | |||
AK316368.1 | - | 6 | 1165 | |||
BC127827.1 | - | 10 | 1480 | 40131713 | ||
BC128591.1 | - | 8 | 1280 | 40127675 | ||
BC146902.1 | - | 5 | 1034 | 9056830 | ||
BC146907.1 | - | 5 | 1034 | 9056835 | ||
DQ232882.1 | - | 5 | 1131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
