siRNA Details
Gene Symbol: PRAMEF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039361.4 | NP_001034450.3 | 3 | 817 | ||
GenBank mRNA | AL049682.1 | - | 3 | 817 | ||
BC112208.1 | - | 3 | 813 | 8327676 | ||
BC112210.1 | - | 3 | 813 | 8327678 |
Gene Symbol: PRAMEF33P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291381.1 | NP_001278310.1 | 3 | 810 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
