siRNA Details
Gene Symbol: SERPINA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006220.2 | NP_006211.2 | 592 | |||
NR_110563.1 | 1 | 592 | ||||
GenBank mRNA | JX680599.1 | - | 588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
