siRNA Details
Gene Symbol: LOC101927051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109952.1 | 2 | 279 | |||
GenBank mRNA | AK097791.1 | - | 3 | 279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
