siRNA Details
Gene Symbol: SERINC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258031.1 | NP_001244960.1 | 12 | 1486 | ||
NM_001258032.1 | NP_001244961.1 | 11 | 1315 | |||
GenBank mRNA | AK097682.1 | - | 10 | 1096 | ||
BC112349.1 | - | 1 | 477 | |||
BC136669.1 | - | 9 | 1111 | 9020659 | ||
BX647438.1 | - | 11 | 1315 | |||
DQ103711.1 | - | 11 | 1275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
