siRNA Details
Gene Symbol: PCDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105243.1 | NP_001098713.1 | 1 | 2938 | ||
NM_001184880.1 | NP_001171809.1 | 1 | 2938 | |||
NM_020766.2 | NP_065817.2 | 1 | 2938 | |||
XM_011530997.2 | XP_011529299.1 | 1 | 1907 | |||
GenBank mRNA | EF676096.1 | - | 1 | 2938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
