siRNA Details
Gene Symbol: PCDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105243.1 | NP_001098713.1 | 5 | 5324 | ||
NM_001184880.1 | NP_001171809.1 | 6 | 5465 | |||
NM_020766.2 | NP_065817.2 | 5 | 5321 | |||
XM_011530997.2 | XP_011529299.1 | 6 | 4431 | |||
GenBank mRNA | AB037734.1 | - | 5 | 1528 | ||
AK096591.1 | - | 5 | 2277 | |||
BC136628.1 | - | 5 | 2205 | 9020618 | ||
CR749278.1 | - | 5 | 2202 | |||
EF676096.1 | - | 6 | 5465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
