siRNA Details
Gene Symbol: SLC6A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010898.2 | NP_001010898.1 | 11 | 2187 | ||
XM_006710643.2 | XP_006710706.1 | 11 | 1895 | |||
GenBank mRNA | AK289982.1 | - | 11 | 2183 | ||
AK297429.1 | - | 2 | 170 | |||
BC140908.1 | - | 10 | 1772 | 9056720 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
