siRNA Details
Gene Symbol: SLC6A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010898.2 | NP_001010898.1 | 12 | 2389 | ||
XM_006710643.2 | XP_006710706.1 | 12 | 2097 | |||
GenBank mRNA | AK289982.1 | - | 12 | 2385 | ||
AK297429.1 | - | 3 | 372 | |||
BC140908.1 | - | 11 | 1974 | 9056720 |

More Information
Other Species Targeted

Gene Ontology Categories:
