siRNA Details
Gene Symbol: LSM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144703.2 | NP_653304.2 | 9 | 2345 | ||
XM_005260302.3 | XP_005260359.1 | 10 | 2457 | |||
XM_011528605.2 | XP_011526907.1 | 11 | 2649 | |||
XM_011528606.2 | XP_011526908.1 | 11 | 2567 | |||
XM_011528607.2 | XP_011526909.1 | 10 | 2575 | |||
XM_011528608.2 | XP_011526910.1 | 10 | 2538 | |||
XM_011528609.2 | XP_011526911.1 | 9 | 2419 | |||
XM_011528611.2 | XP_011526913.1 | 8 | 2263 | |||
XM_011528612.2 | XP_011526914.1 | 11 | 2319 | |||
XM_011528613.2 | XP_011526915.1 | 10 | 2179 | |||
XM_011528614.2 | XP_011526916.1 | 10 | 2392 | |||
XM_011528615.2 | XP_011526917.1 | 7 | 1876 | |||
XM_017027688.1 | XP_016883177.1 | 8 | 2252 | |||
XM_017027689.1 | XP_016883178.1 | 8 | 2171 | |||
XM_017027690.1 | XP_016883179.1 | 8 | 1993 | |||
GenBank mRNA | BC054888.1 | - | 1 | 335 | ||
CX166280.1 | - | 1 | 411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
