siRNA Details
Gene Symbol: OVOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020288.1 | XP_016875777.1 | 28 | 3891 | ||
GenBank mRNA | CR749395.1 | - | 2 | 1559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
