siRNA Details
Gene Symbol: FAM135B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015912.3 | NP_056996.2 | 19, 20 | 4194 | ||
XM_011517056.2 | XP_011515358.1 | 20, 21 | 5012 | |||
XM_011517058.2 | XP_011515360.1 | 19, 20 | 4867 | |||
XM_011517059.1 | XP_011515361.1 | 19, 20 | 4134 | |||
XM_011517060.2 | XP_011515362.1 | 20, 21 | 4994 | |||
XM_011517061.2 | XP_011515363.1 | 20, 21 | 4351 | |||
XM_011517062.1 | XP_011515364.1 | 19, 20 | 4206 | |||
XM_011517063.2 | XP_011515365.1 | 19, 20 | 4849 | |||
XM_011517064.2 | XP_011515366.1 | 20, 21 | 4999 | |||
XM_011517065.1 | XP_011515367.1 | 19, 20 | 4270 | |||
XM_011517068.2 | XP_011515370.1 | 19, 20 | 4087 | |||
XM_011517069.2 | XP_011515371.1 | 14, 15 | 4193 | |||
XM_011517072.2 | XP_011515374.1 | 13, 14 | 3451 | |||
XM_011517073.2 | XP_011515375.1 | 13, 14 | 3455 | |||
XM_011517074.1 | XP_011515376.1 | 12, 13 | 3321 | |||
XM_017013470.1 | XP_016868959.1 | 19, 20 | 4806 | |||
XM_017013471.1 | XP_016868960.1 | 20, 21 | 4955 | |||
GenBank mRNA | AB196635.1 | - | 19, 20 | 4091 | ||
BC037288.1 | - | 20, 21 | 4229 | 5266568 | ||
BX110118.1 | - | 1, 2 | 53 | |||
CR936831.1 | - | 13, 14 | 3387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
