siRNA Details
Gene Symbol: LINC00266-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040415.1 | 3 | 583 | |||
GenBank mRNA | BC118988.1 | - | 3 | 583 | 40035995 | |
BC122537.1 | - | 3 | 583 | 40035996 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
