siRNA Details
Gene Symbol: AKR1C6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026743.1 | 5 | 477 | |||
GenBank mRNA | AB037902.1 | - | 3 | 280 | ||
CB111913.1 | - | 5 | 432 | |||
DA643129.1 | - | 4 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
