siRNA Details
Gene Symbol: SERP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010897.1 | NP_001010897.1 | 3 | 478 | ||
GenBank mRNA | AK123212.1 | - | 1 | 1278 | ||
BC029067.1 | - | 3 | 478 | 5197847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
