siRNA Details
Gene Symbol: CCDC175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164399.1 | NP_001157871.1 | 3 | 351 | ||
XM_011537120.2 | XP_011535422.1 | 3 | 788 | |||
XM_011537122.2 | XP_011535424.1 | 3 | 787 | |||
XM_011537124.2 | XP_011535426.1 | 3 | 787 | |||
XM_011537125.2 | XP_011535427.1 | 3 | 792 | |||
XM_011537130.2 | XP_011535432.1 | 3 | 790 | |||
XM_017021621.1 | XP_016877110.1 | 3 | 790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
